Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhx32
Entrez Gene ID
361667
Gene Name
DEAH (Asp-Glu-Ala-His) box polypeptide 32
Gene Aliases
Ddx32
Chromosome Location
Chr.1: 205789442 - 205842428 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130039.1 NP_001123511.1 4 - 5 1265 112
XM_006230443.2 XP_006230505.1 4 - 5 1830 112

View Full Product Details