Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tollip
Entrez Gene ID
361677
Gene Name
toll interacting protein
Gene Aliases
-
Chromosome Location
Chr.1: 214823993 - 214844861 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109668.1 NP_001103138.1 3 - 4 565 107
XM_006230607.2 XP_006230669.1 4 - 5 646 107
XM_008760003.1 XP_008758225.1 3 - 4 510 107
GenBank mRNA BC133067.1 - 3 - 4 451 107

View Full Product Details