Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Actn4
Entrez Gene ID
63836
Gene Name
actinin alpha 4
Gene Aliases
-
Chromosome Location
Chr.1: 87078012 - 87147347 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031675.2 NP_113863.2 18 - 19 2447 109
XM_006228687.1 XP_006228749.1 18 - 19 2486 109
XM_006228690.2 XP_006228752.1 18 - 19 2765 109
XM_008759185.1 XP_008757407.1 18 - 19 2486 109
GenBank mRNA AF190909.1 - 18 - 19 2372 109
AF336827.1 - 9 - 10 1241 109
AY724534.1 - 18 - 19 2416 109
BC061788.1 - 18 - 19 2438 109
U19893.1 - 18 - 19 2448 109

View Full Product Details