Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Six5
Entrez Gene ID
308406
Gene Name
SIX homeobox 5
Gene Aliases
-
Chromosome Location
Chr.1: 80000119 - 80005303 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003748780.3 XP_003748828.2 2 - 3 1619 87

View Full Product Details