The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008292.1 | NP_001008293.1 | 5 - 6 | 538 | 62 |
GenBank mRNA | BC086366.1 | - | 5 - 6 | 538 | 62 |
FQ219826.1 | - | 5 - 6 | 531 | 62 |