Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sppl3
Entrez Gene ID
360822
Gene Name
signal peptide peptidase like 3
Gene Aliases
RGD1562826, Sppl3-ps1
Chromosome Location
Chr.12: 47276485 - 47360552 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751200.3 XP_003751248.1 10 - 11 1187 105

View Full Product Details