The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053760.2 | NP_446212.2 | 6 - 7 | 950 | 60 |
XM_006249376.2 | XP_006249438.1 | 6 - 7 | 1006 | 60 | |
XM_008769209.1 | XP_008767431.1 | 6 - 7 | 1053 | 60 |