Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trafd1
Entrez Gene ID
114635
Gene Name
TRAF type zinc finger domain containing 1
Gene Aliases
Fln29
Chromosome Location
Chr.12: 40695417 - 40709438 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053760.2 NP_446212.2 8 - 9 1256 75
XM_006249376.2 XP_006249438.1 8 - 9 1312 75
XM_008769209.1 XP_008767431.1 8 - 9 1359 75
GenBank mRNA AF329825.1 - 2 - 3 260 75

View Full Product Details