The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053626.1 | NP_446078.1 | 9 - 10 | 1003 | 83 |
GenBank mRNA | AB003400.1 | - | 9 - 10 | 1003 | 83 |
BC088395.1 | - | 8 - 9 | 918 | 83 |