Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh2b3
Entrez Gene ID
58838
Gene Name
SH2B adaptor protein 3
Gene Aliases
Lnk
Chromosome Location
Chr.12: 40261990 - 40265757 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JX215366.1 - - 981 100

View Full Product Details