The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105919.1 | NP_001099389.1 | 4 - 5 | 633 | 72 |
XM_006249123.2 | XP_006249185.1 | 4 - 5 | 679 | 72 | |
GenBank mRNA | FQ213833.1 | - | 4 - 5 | 459 | 72 |
FQ222246.1 | - | 4 - 5 | 445 | 72 | |
FQ224250.1 | - | 4 - 5 | 456 | 72 |