Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wbscr17
Entrez Gene ID
288611
Gene Name
Williams-Beuren syndrome chromosome region 17
Gene Aliases
-
Chromosome Location
Chr.12: 28382331 - 29268453 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025112.1 NP_001020283.1 4 - 5 761 72
XM_008769132.1 XP_008767354.1 4 - 5 629 72
GenBank mRNA AB040671.1 - 4 - 5 761 72

View Full Product Details