Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sh2b2
Entrez Gene ID
114203
Gene Name
SH2B adaptor protein 2
Gene Aliases
Aps
Chromosome Location
Chr.12: 23471244 - 23501043 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053669.1 NP_446121.1 5 - 6 1269 124
XM_006249092.2 XP_006249154.1 5 - 6 2021 124
XM_008769112.1 XP_008767334.1 6 - 7 1820 124
XM_008769113.1 XP_008767335.1 5 - 6 1284 124
GenBank mRNA AF095576.1 - 5 - 6 1269 124

View Full Product Details