The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022512.2 | NP_071957.1 | 9 - 10 | 1118 | 119 |
GenBank mRNA | BC072545.1 | - | 9 - 10 | 1137 | 119 |
FQ227562.1 | - | 3 - 4 | 289 | 119 | |
J05030.1 | - | 9 - 10 | 1119 | 119 |