Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Caln1
Entrez Gene ID
363909
Gene Name
calneuron 1
Gene Aliases
-
Chromosome Location
Chr.12: 29307117 - 29743705 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077201.1 NP_001070669.1 2 - 3 240 119
XM_006249243.2 XP_006249305.1 2 - 3 207 119
GenBank mRNA DQ914829.1 - 1 - 2 114 119
DQ914830.1 - 2 - 3 240 119

View Full Product Details