The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033675.1 | NP_001028847.1 | 4 - 5 | 337 | 98 |
NM_001033676.1 | NP_001028848.1 | 4 - 5 | 949 | 98 | |
NM_133529.2 | NP_598213.1 | 5 - 6 | 517 | 98 | |
GenBank mRNA | AJ315657.1 | - | 5 - 6 | 517 | 98 |
AJ315761.1 | - | 4 - 5 | 337 | 98 | |
Y17048.2 | - | 4 - 5 | 949 | 98 |