Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Carkd
Entrez Gene ID
361185
Gene Name
carbohydrate kinase domain containing
Gene Aliases
RGD1562691
Chromosome Location
Chr.16: 83326228 - 83342688 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108402.1 NP_001101872.1 5 - 6 453 151
XM_006253465.2 XP_006253527.1 5 - 6 580 151
XM_008771403.1 XP_008769625.1 5 - 6 471 151
GenBank mRNA FQ225763.1 - - 415 151

View Full Product Details