Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ido1
Entrez Gene ID
66029
Gene Name
indoleamine 2,3-dioxygenase 1
Gene Aliases
Ido, Indo
Chromosome Location
Chr.16: 72216326 - 72228098 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023973.1 NP_076463.1 9 - 10 872 82
GenBank mRNA AF312699.1 - 9 - 10 872 82

View Full Product Details