Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kbtbd11
Entrez Gene ID
306617
Gene Name
kelch repeat and BTB (POZ) domain containing 11
Gene Aliases
RGD1563576
Chromosome Location
Chr.16: 79696395 - 79716155 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107326.1 NP_001100796.1 1 - 1 3849 122
XM_006253390.2 XP_006253452.1 2 - 2 4579 122
XM_006253391.2 XP_006253453.1 2 - 2 4574 122
XM_006253392.2 XP_006253454.1 2 - 2 4417 122
XM_006253393.2 XP_006253455.1 2 - 2 4457 122

View Full Product Details