Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grtp1
Entrez Gene ID
361180
Gene Name
growth hormone regulated TBC protein 1
Gene Aliases
LRRGT00052
Chromosome Location
Chr.16: 81665533 - 81687845 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170477.1 NP_001163948.1 3 - 4 437 76

View Full Product Details