The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031096.1 | NP_112358.1 | 5 - 6 | 1384 | 138 |
GenBank mRNA | U63971.1 | - | 5 - 6 | 1384 | 138 |