Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ido2
Entrez Gene ID
681319
Gene Name
indoleamine 2,3-dioxygenase 2
Gene Aliases
Indol1
Chromosome Location
Chr.16: 72232852 - 72291227 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771375.1 XP_008769597.1 3 - 4 188 125

View Full Product Details