Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zc3h11a
Entrez Gene ID
360845
Gene Name
zinc finger CCCH-type containing 11A
Gene Aliases
RGD1308290
Chromosome Location
Chr.13: 50196042 - 50236541 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047902.1 NP_001041367.1 12 - 13 1780 69
XM_008769482.1 XP_008767704.1 10 - 11 1557 69
XM_008769483.1 XP_008767705.1 10 - 11 1554 69
XM_008769484.1 XP_008767706.1 10 - 11 1530 69
GenBank mRNA BC111404.1 - 5 - 6 793 69
DQ480752.1 - 12 - 13 1780 69

View Full Product Details