Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zc3h11a
Entrez Gene ID
360845
Gene Name
zinc finger CCCH-type containing 11A
Gene Aliases
RGD1308290
Chromosome Location
Chr.13: 50196042 - 50236541 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047902.1 NP_001041367.1 7 - 8 933 112
XM_008769482.1 XP_008767704.1 5 - 6 710 112
GenBank mRNA DQ480752.1 - 7 - 8 933 112

View Full Product Details