Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubxn4
Entrez Gene ID
304766
Gene Name
UBX domain protein 4
Gene Aliases
Ubxd2
Chromosome Location
Chr.13: 44956963 - 44989653 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012025.1 NP_001012025.1 11 - 12 1471 127
XM_006249699.2 XP_006249761.1 11 - 12 1517 127
GenBank mRNA BC088845.1 - 11 - 12 1471 127

View Full Product Details