Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lhx9
Entrez Gene ID
289048
Gene Name
LIM homeobox 9
Gene Aliases
-
Chromosome Location
Chr.13: 55854543 - 55879305 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181367.2 NP_852032.1 1 - 2 85 55
XM_006249916.2 XP_006249978.1 2 - 3 647 55
XM_006249919.2 XP_006249981.1 2 - 3 647 55
XM_008769545.1 XP_008767767.1 1 - 2 173 55
GenBank mRNA AY273890.1 - 1 - 2 85 55
AY724502.1 - 1 - 2 85 55
BC128722.1 - 2 - 3 161 55

View Full Product Details