Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pamr1
Entrez Gene ID
311252
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
RGD1308745, Ramp
Chromosome Location
Chr.3: 92403532 - 92502094 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107755.1 NP_001101225.1 5 - 6 772 75
XM_008762061.1 XP_008760283.1 5 - 6 889 75

View Full Product Details