Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Accsl
Entrez Gene ID
690470
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (Arabidopsis)(non-functional)-like
Gene Aliases
-
Chromosome Location
Chr.3: 82762861 - 82777814 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109594.1 NP_001103064.1 4 - 5 748 84
XM_008762035.1 XP_008760257.1 3 - 4 776 84

View Full Product Details