Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmco5a
Entrez Gene ID
499865
Gene Name
transmembrane and coiled-coil domains 5A
Gene Aliases
RGD1565206, Tmco5
Chromosome Location
Chr.3: 108544916 - 108559448 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006234788.2 XP_006234850.1 3 - 4 304 80
XM_006234789.2 XP_006234851.1 3 - 4 284 80
XM_006234790.2 XP_006234852.1 3 - 4 329 80
XM_006234791.2 XP_006234853.1 2 - 3 186 80
XM_008762112.1 XP_008760334.1 3 - 4 361 80

View Full Product Details