Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ell3
Entrez Gene ID
296102
Gene Name
elongation factor RNA polymerase II-like 3
Gene Aliases
-
Chromosome Location
Chr.3: 113400474 - 113405861 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011957.1 NP_001011957.1 9 - 10 1065 89
GenBank mRNA BC084693.1 - 9 - 10 1065 89

View Full Product Details