Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGD1564664
Entrez Gene ID
499839
Gene Name
similar to LOC387763 protein
Gene Aliases
Ag2
Chromosome Location
Chr.3: 82854779 - 82856171 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110055.1 NP_001103525.1 1 - 1 929 147
GenBank mRNA AB010079.1 - 1 - 1 929 147

View Full Product Details