Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phospho2
Entrez Gene ID
295663
Gene Name
phosphatase, orphan 2
Gene Aliases
-
Chromosome Location
Chr.3: 56030915 - 56038076 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007642.1 NP_001007643.1 2 - 3 200 85
GenBank mRNA BC081926.1 - 2 - 3 200 85

View Full Product Details