The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106487.1 | NP_001099957.1 | 2 - 3 | 448 | 67 |
XM_006234449.1 | XP_006234511.1 | 3 - 4 | 484 | 67 | |
XM_006234450.1 | XP_006234512.1 | 3 - 4 | 467 | 67 |