Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mdk
Entrez Gene ID
81517
Gene Name
midkine
Gene Aliases
-
Chromosome Location
Chr.3: 80841003 - 80843913 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030859.2 NP_110486.1 2 - 3 233 81
XM_006234591.2 XP_006234653.1 3 - 4 398 81
XM_006234592.2 XP_006234654.1 3 - 4 346 81
XM_006234593.2 XP_006234655.1 3 - 4 1023 81
GenBank mRNA AB025023.1 - 2 - 3 233 81
AF315950.1 - 3 - 4 312 81
BC157805.1 - 3 - 4 264 81

View Full Product Details