Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ripply1
Entrez Gene ID
680702
Gene Name
ripply transcriptional repressor 1
Gene Aliases
-
Chromosome Location
Chr.X: 111099420 - 111102539 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008773499.1 XP_008771721.1 2 - 3 308 62

View Full Product Details