Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rapsn
Entrez Gene ID
362161
Gene Name
receptor-associated protein of the synapse
Gene Aliases
-
Chromosome Location
Chr.3: 79859816 - 79869486 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006234528.2 XP_006234590.1 3 - 4 1244 55

View Full Product Details