Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rapsn
Entrez Gene ID
362161
Gene Name
receptor-associated protein of the synapse
Gene Aliases
-
Chromosome Location
Chr.3: 79859816 - 79869486 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108584.1 NP_001102054.1 3 - 4 794 77
XM_006234528.2 XP_006234590.1 4 - 5 1344 77

View Full Product Details