Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mtmr2
Entrez Gene ID
315422
Gene Name
myotubularin related protein 2
Gene Aliases
-
Chromosome Location
Chr.8: 12283436 - 12334014 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108123.1 NP_001101593.1 14 - 15 2764 142
XM_006242521.2 XP_006242583.1 15 - 16 3074 142
XM_006242522.1 XP_006242584.1 14 - 15 1891 142
XM_008765932.1 XP_008764154.1 15 - 16 1781 142

View Full Product Details