Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casp12
Entrez Gene ID
156117
Gene Name
caspase 12
Gene Aliases
-
Chromosome Location
Chr.8: 2658801 - 2689184 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130422.1 NP_569106.1 5 - 6 684 118
XM_008765891.1 XP_008764113.1 6 - 7 765 118
GenBank mRNA AF317633.1 - 5 - 6 684 118
FJ465154.1 - 6 - 7 747 118

View Full Product Details