Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atg4d
Entrez Gene ID
686505
Gene Name
autophagy related 4D, cysteine peptidase
Gene Aliases
-
Chromosome Location
Chr.8: 22291445 - 22301331 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101013.2 NP_001094483.2 3 - 4 682 71
GenBank mRNA BC168141.1 - 3 - 4 404 71

View Full Product Details