Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmp27
Entrez Gene ID
300340
Gene Name
matrix metallopeptidase 27
Gene Aliases
-
Chromosome Location
Chr.8: 5790019 - 5799938 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106799.1 NP_001100269.1 4 - 5 663 76

View Full Product Details