The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138878.2 | NP_620233.1 | 3 - 4 | 405 | 84 |
GenBank mRNA | AF095740.1 | - | 3 - 4 | 204 | 84 |
BC084728.1 | - | 3 - 4 | 225 | 84 | |
CB581170.1 | - | 3 - 4 | 416 | 84 | |
FQ230524.1 | - | 2 - 3 | 131 | 84 |