Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rcbtb1
Entrez Gene ID
361050
Gene Name
regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 1
Gene Aliases
-
Chromosome Location
Chr.15: 39622572 - 39665335 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108380.1 NP_001101850.1 1 - 2 325 73

View Full Product Details