Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cideb
Entrez Gene ID
364388
Gene Name
cell death-inducing DFFA-like effector b
Gene Aliases
-
Chromosome Location
Chr.15: 34439844 - 34444244 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006252002.2 XP_006252064.1 2 - 3 352 105

View Full Product Details