Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jph4
Entrez Gene ID
445271
Gene Name
junctophilin 4
Gene Aliases
-
Chromosome Location
Chr.15: 33767116 - 33776447 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003711.1 NP_001003711.1 2 - 3 1158 62
XM_008770690.1 XP_008768912.1 3 - 4 1956 62
GenBank mRNA AY341260.1 - 2 - 3 1158 62

View Full Product Details