The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106411.1 | NP_001099881.1 | 1 - 2 | 238 | 74 |
GenBank mRNA | FQ214900.1 | - | 1 - 2 | 212 | 74 |
FQ220265.1 | - | 1 - 2 | 218 | 74 | |
FQ220543.1 | - | 1 - 2 | 218 | 74 | |
FQ229597.1 | - | 1 - 2 | 212 | 74 | |
FQ229607.1 | - | 1 - 2 | 208 | 74 | |
FQ229635.1 | - | 1 - 2 | 212 | 74 | |
FQ229721.1 | - | 1 - 2 | 212 | 74 | |
FQ230033.1 | - | 1 - 2 | 212 | 74 |