Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mier3
Entrez Gene ID
310086
Gene Name
mesoderm induction early response 1, family member 3
Gene Aliases
RGD1311241
Chromosome Location
Chr.2: 43266316 - 43295175 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168000.1 NP_001161472.1 9 - 10 1017 120
XM_006231924.2 XP_006231986.1 11 - 12 1112 120
XM_006231925.2 XP_006231987.2 11 - 12 1519 120
GenBank mRNA JQ013732.1 - 9 - 10 1963 120

View Full Product Details