Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pm20d1
Entrez Gene ID
498226
Gene Name
peptidase M20 domain containing 1
Gene Aliases
RGD1564063
Chromosome Location
Chr.13: 48575094 - 48596515 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109068.1 NP_001102538.1 2 - 3 292 64

View Full Product Details