Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vamp4
Entrez Gene ID
364033
Gene Name
vesicle-associated membrane protein 4
Gene Aliases
-
Chromosome Location
Chr.13: 80460699 - 80483597 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108856.1 NP_001102326.1 3 - 4 233 136
XM_006250184.2 XP_006250246.1 3 - 4 320 136
XM_006250186.2 XP_006250248.1 3 - 4 312 136

View Full Product Details