Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ren
Entrez Gene ID
24715
Gene Name
renin
Gene Aliases
RATRENAA, RENAA, Ren1
Chromosome Location
Chr.13: 50502724 - 50513953 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF117820.1 - 1 - 2 44 75
AF233692.1 - 1 - 2 77 75

View Full Product Details